Sphingosine-1-phosphate ≥95%, Calbiochem®
Supplier: MilliporeSigma
Synonyms:
S1P
, D-erythro-Sphingosine 1-phosphate
Packaged under inert gas.
567727-1MG
80054-486EA
234.69
USD
80054-486
Sphingosine-1-phosphate ≥95%, Calbiochem®
Sphingosine-1-phosphate
A putative second messenger that mobilizes calcium from intracellular stores via an IP3-independent pathway.
Activates phospholipase D (PLD) and inhibits chemotactic motility and invasiveness of tumor cells. Also stimulates growth of quiescent Swiss/3T3 fibroblasts. Inhibits activation of caspases that cleave PARP and lamins during Fas- and ceramide-mediated apoptosis.
Formula:
C₁₈H₃₈NO₅P MW: 379.478 g/mol |
MDL Number:
MFCD00270077 CAS Number: 26993-30-6 |
Specification Test Results
Form | White Solid |
Chemical formula | C18H38NO5P |
Hygroscopic | Hygroscopic |
Reversible | N |
Primary Target | Mobilizes calcium from intracellular stores |
Purity | ≥95% by TLC |
Cell Permeable | N |
Toxicity | Irritant |
Storage | -20 °C |
Hygroscopic | Hygroscopic |
Do not Freeze | Ok to Freeze |
Special Instructions | Reconstitution, aliquot and evaporate under a stream of nitrogen |
Usage | Prior to use, dissolve in 4 mg/ml BSA solution with vortexing |
Life | Stock solutions are stable for up to 2 months at -20°C |
Package | Packaged under Inert gas |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...