Diethyl azodicarboxylate 40% in toluene
Supplier: Pfaltz & Bauer
|
Danger
|
D21100-25G
101188-478EA
463.38
USD
101188-478
Diethyl azodicarboxylate 40% in toluene
Diethyl azodicarboxylate
Formula:
C₆H₁₀N₂O₄ Boiling Pt: 116…117 °C Density: 0.956 Flash Pt: 41 °C |
MDL Number:
MFCD00009103 CAS Number: 1972-28-7 UN: 3223 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...