4-Acetoxy-3-methoxyphenylacetic acid ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

195761GM
100267-546EA 85 USD
100267-546
4-Acetoxy-3-methoxyphenylacetic acid ≥98%, powder
4-Acetoxy-3-methoxyphenylacetic acid

  • White powder
  • Store in a cool, dry place

Melting Pt: 138…140 °C
MDL Number: MFCD03424354
CAS Number: 5447-38-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/62/21285762.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR