Polyethylene terephthalate

Supplier: USP

Synonyms: 1,4-Benzenedicarboxylic acid, polymer with 1,2-ethanediolArnite AAmilar

1546900
101095-858EA 424.62 USD
101095-858
Polyethylene terephthalate
Polyethylene terephthalate

  • 3 Strips 2'' × 2 1/2''

Formula: (C₁₀H₈O₄)n
MDL Number: MFCD00084422
CAS Number: 25038-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR