Would you like to log in again?
Aroclors Solutions and Kits, Ultra Scientific
Supplier: ULTRA SCIENTIFIC
1242TK 1232TK 1254TK 1221TK 1262TK 1016TK 1260TK-B2 1260TK 1248TK-B2 1248TK-B1 1248TK 1242TK-B1 1242TK-B2 1268TK 1254TK-B1 1254TK-B2
Industrial PCBs were generally prepared by treating biphenyl with anhydrous chlorine in the presence of a catalyst (iron filings or ferric chloride).
Delivery information: Each kit contains four ampules of 3 x 2 mL of Aroclor, 1 x 2 mL of PCB Free Transformer Oil, Commercial PCBs.
ORDER
Expand All / Collapse All
Kits
Description Concentration Supplier No. VWR Catalog Number Unit Price Quantity
Aroclor 1016 Kit 100 µg/mL in transformer oil (PCB-free) 1016TK UL1016TK Each Retrieving Restricted
Small Business Enterprise
Aroclor 1221 Kit 100 µg/mL in transformer oil (PCB-free) 1221TK UL1221TK Each Retrieving Restricted
Small Business Enterprise
Aroclor 1232 Kit 100 µg/mL in transformer oil (PCB-free) 1232TK UL1232TK Each Retrieving Restricted
Small Business Enterprise
Aroclor 1242 Kit 100 µg/mL in transformer oil (PCB-free) 1242TK UL1242TK Each Retrieving Restricted
Small Business Enterprise
Aroclor 1248 Kit 100 µg/mL in transformer oil (PCB-free) 1248TK UL1248TK Each Retrieving Restricted
Small Business Enterprise
Aroclor 1254 Kit 100 µg/mL in transformer oil (PCB-free) 1254TK UL1254TK Each Retrieving Restricted
Small Business Enterprise
Aroclor 1260 Kit 100 µg/mL in transformer oil (PCB-free) 1260TK UL1260TK Each Retrieving Restricted
Small Business Enterprise
Aroclor 1262 Kit 100 µg/mL in transformer oil (PCB-free) 1262TK UL1262TK Each Retrieving Restricted
Small Business Enterprise
View MSDS SDS
Aroclor 1268 Kit 100 µg/mL in transformer oil (PCB-free) 1268TK UL1268TK Each Retrieving Restricted
Small Business Enterprise
View MSDS SDS
Solutions
Description Size Concentration Supplier No. VWR Catalog Number Unit Price Quantity
Aroclor 1242 Solution 1 x 1 mL 500 µg/mL in transformer oil (PCB-free) 1242TK-B2 UL1242TK-B2 Each (1ml) Retrieving Restricted
Small Business Enterprise
Aroclor 1242 Solution 1 x 1 mL 50 µg/mL in transformer oil (PCB-free) 1242TK-B1 UL1242TK-B1 Each (1ml) Retrieving Restricted
Small Business Enterprise
Aroclor 1248 Solution 1 x 1 mL 50 µg/gm in transformer oil (PCB-free) 1248TK-B1 UL1248TK-B1 Each (1ml) Retrieving Restricted
Small Business Enterprise
Aroclor 1248 Solution 1 x 1 mL 500 µg/gm in transformer oil (PCB-free) 1248TK-B2 UL1248TK-B2 Each (1ml) Retrieving Restricted
Small Business Enterprise
Aroclor 1254 Solution 1 x 1 mL 500 µg/gm in transformer oil (PCB-free) 1254TK-B2 UL1254TK-B2 Each (1ml) Retrieving Restricted
Small Business Enterprise
Aroclor 1254 Solution 1 x 1 mL 50 µg/gm in transformer oil (PCB-free) 1254TK-B1 UL1254TK-B1 Each (1ml) Retrieving Restricted
Small Business Enterprise
Aroclor 1260 Solution 1 x 1 mL 500 µg/gm in transformer oil (PCB-free) 1260TK-B2 UL1260TK-B2 Each (1ml) Retrieving Restricted
Small Business Enterprise
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty