Fluorescein diacetate, Calbiochem®

Supplier: MilliporeSigma

343209-5GM
80109-168EA 167.4 USD
80109-168
Fluorescein diacetate, Calbiochem®
Fluorescein diacetate

Stains living cells green. Substrate for esterases. Suitable for use in cell viability assays. Removal of the acetate groups by cellular esterases results in fluorescence in cells with intact membranes.

Formula: C₂₄H₁₆O₇
Storage Temperature: Refrigerator
MDL Number: MFCD00005062
CAS Number: 596-09-8

Order Now

Specification Test Results

Storage 2 - 8 °C, Protect from light
Special Instructions Unstable in solution. Reconstitute just prior to use.
Appearance Colorless to Yellow Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR