Difco™ BG Sulfa Agar, BD
Supplier: BD Life Sciences
271710 271710
90003-882EA 518.9 USD
90003-882 CA90003-882
Difco™ BG Sulfa Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
BG Sulfa Agar is a highly selective medium for the isolation of Salmonella.
  • Peptone and yeast extract provide nitrogen, vitamins, and minerals
  • Lactose and sucrose are the sources of carbohydrates
  • Phenol red acts as a pH indicator
Brilliant green and sodium pyridine are complementary in inhibiting gram-positive bacteria and most gram-negative bacilli other than Salmonella species.