BBL™ AK Agar #2 (Sporulating Agar), BD
Supplier: BD Biosciences
210912 210912
90000-014EA 496.43 USD
CA90000-014 90000-014
BBL™ AK Agar #2 (Sporulating Agar), BD
Media for Microbiology Dehydrated Media for Microbiology
AK Agar #2 (Sporulating Agar) is a culture medium for the preparation of spore suspensions for use in procedures for the detection of antibiotic residues in milk and dairy products.
  • Yeast extract provides B vitamins
  • Dextrose is an energy source for bacterial replication
  • Manganous sulfate plays an important role in the sporulation process
Peptones and beef extract are sources of nitrogen, sulfur, amino acids, and essential trace ingredients.