Difco™ Pepsin 1:10,000, BD Diagnostics
Supplier: BD Diagnostics
215110
90000-892EA 658.69 USD
90000-892
Difco™ Pepsin 1:10,000, BD Diagnostics
Pepsin is an enzyme that breaks down proteins into smaller peptides.
  • Most active in acidic environments between 37°C and 42°C
  • Maximal activity at pH 2.0
  • Inactive at pH 6.5 and above
  • Fully denatured at pH 8.0
Pepsin is produced in the stomach and is one of the main digestive enzymes in the digestive systems of humans and many other animals, where it helps digest the proteins in food.