Tronex® Procedure Face Mask with Earloops, Tronex International, Inc.
Supplier: Tronex
Tronex®
5040B
10152-984BX 4.19 USD
10152-984
Tronex® Procedure Face Mask with Earloops, Tronex International, Inc.
Masks Hygiene Masks
Quality crafted from soft, breathable material for excellent protection and a comfortable fit.
  • ≥98% Bacterial and Particulate Filtration Efficiency (BFE/PFE)
  • Constructed with ultrasonic bonding for enhanced durability
  • Malleable nosepiece contours are comfortable to all facial shapes
  • Elastic earloops foster easy donning and a secure fit
  • 100% latex-free
Ideal for applications where fluid exposure is probable.
Specialized nonwoven material structure features three-ply, pleated construction to optimize lightness of fit, breathability, and fluid resistance.
Certifications: Manufacturing certified ISO 9001 for Quality Management Systems and ISO 13485 for Quality Management Systems for Medical Devices.