Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Jacketed Indented Cylindrical Reaction Vessels with Flat Flange, Wilmad-LabGlass

Supplier: WILMAD/LABGLASS
LG-8085-108 LG-8085-106 LG-8085-104 LG-8085-102 LG-8085-100
These jacketed, cylindrical borosilicate glass reaction vessels have four indentations for improved stirring.

  • Sturdy walls
  • Tooled flat flange

They are without constriction at the top and have a flat bottom.They feature sturdy walls and a tooled flat flange with a surface ground to assure a tight seal. Jacket has 1/2" hose connectors.

Ordering information: Uses LG-7316 clamps.



 

ORDER

Expand All / Collapse All
Capacity For Use With Body Diameter Overall Height Flange O.D. Supplier No. VWR Catalog Number Unit Price Quantity
500 mL Clamp LG-7316-106 3-3/4" 6" 5-3/8" LG-8085-100 14205-944 Each Retrieving
1000 mL Clamp LG-7316-106 4-1/4" 6 1/2" 5-3/8" LG-8085-102 14205-250 Each Retrieving
2000 mL Clamp LG-7316-108 5-1/2" 7 1/8" 6-5/8" LG-8085-104 14205-538 Each Retrieving
3000 mL Clamp LG-7316-108 5-1/2" 10 1/8" 6-5/8" LG-8085-106 14207-482 Each Retrieving
4000 mL Clamp LG-7316-108 5-1/2" 13 1/4" 6-5/8" LG-8085-108 14206-092 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty