Would you like to log in again?
AlphaGuard® Scrub Shirts and Pants
Supplier: ALPHA PROTECH
WP-23322-2 WS-23221-1 WP-23322-1 WP-23322-4 WS-23221-3 WP-23322-3 WS-23221-2 WP-23322-6 WS-23221-5 WP-23322-5 WS-23221-4 WS-23221-7 WP-23322-7
Scrubs provide a discreet barrier for employees and are highly breathable to keep users cool and dry.
  • Safety stitched seams for added strength and durability
AlphaGuard material is extremely soft and comfortable to wear and delivers high filtration efficiency while maintaining a low particle count. Scrub shirts are available in long sleeve style with a round banded neck, one pocket, and elastic wrist. Scrub pants include a spandex waist, a rear pocket, and are reinforced along the inner thigh area.
ORDER
Expand All / Collapse All
Size Description Supplier No. VWR Catalog Number Unit Price Quantity
Small Pants, Spandex Waist, 1 Pocket WP-23322-1 89126-220 Case of 30 Retrieving
Medium Pants, Spandex Waist, 1 Pocket WP-23322-2 89126-222 Case of 30 Retrieving
Large Pants, Spandex Waist, 1 Pocket WP-23322-3 89126-224 Case of 30 Retrieving
X-Large Pants, Spandex Waist, 1 Pocket WP-23322-4 89126-226 Case of 30 Retrieving
2X-Large Pants, Spandex Waist, 1 Pocket WP-23322-5 89126-228 Case of 30 Retrieving
Small Shirts, Long Sleeve, Round Neck WS-23221-1 89130-798 Case of 30 Retrieving
Medium Shirts, Long Sleeve, Round Neck WS-23221-2 89130-800 Case of 30 Retrieving
Large Shirts, Long Sleeve, Round Neck WS-23221-3 89130-802 Case of 30 Retrieving
X-Large Shirts, Long Sleeve, Round Neck WS-23221-4 89130-804 Case of 30 Retrieving
2X-Large Shirts, Long Sleeve, Round Neck WS-23221-5 89130-806 Case of 30 Retrieving
4X-Large Shirts, Long Sleeve, Round Neck WS-23221-7 89130-810 Case of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty