Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EZDetect™ Aldo-keto Reductase Activity Assay Kit (Colorimetric), BioVision Inc.

Supplier: BioVision
K847-100
Enzymatic AKR activity assay.

  • Simple and Ultrasensitive assay
  • Colorimetrically measure as low as 0.001 mU of AKR activity
  • Provides a convenient tool for sensitive detection of AKR activity in a variety of samples

Aldo-keto Reductases (AKRs) are a superfamily of NAD(P)H linked oxidoreductases which contains more than 190 members. A complete listing of AKR superfamily can be found at: https://www.med.upenn.edu/akr/. The common function of AKR superfamily is to oxidize/reduce aldehydes or ketones to their corresponding primary or secondary alcohols. Due to their broad substrate specificity, AKRs play roles in intermediary metabolism, biosynthesis and detoxification. In humans, the AKRs are not only involved in carbonyl metabolism, but they also regulate signaling through nuclear receptors which leads to chemical carcinogenesis. Many studies demonstrate increasing expression of AKRs in cancers such as lung, liver and colon cancers. BioVision’s EZDetect™ Aldo-keto Reductase Activity Assay Kit provides a convenient tool for sensitive detection of AKR activity in a variety of samples. In this assay, AKR acts on a general substrate and converts NADP+ to NADPH. This NADPH reacts with the AKR probe and generates color. This color is proportional to the activity of AKR in the sample. This kit can detect AKR activity as low as 0.001 mU in samples and has been validated with AKR1B10, AKR1C1 and AKR1C3. Further tests are in progress for other members of the AKR family.




 

ORDER

Expand All / Collapse All
Description Size Storage Temperature Supplier No. VWR Catalog Number Unit Price Quantity
EZDetect™ Aldo-keto Reductase Activity Assay Kit (Colorimetric) 100 assays -20°C K847-100 75832-902 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Species Reactivity Mammalian
Sample Type Animal tissues such as liver, pancreas, small intestine, etc.; Adherent and suspension cells; Biological fluids such as serum and plasma.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty