Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HIV-2 Protease Activity Assay Kit (Fluorometric), BioVision Inc.

Supplier: BioVision
K845-100
Sensitive fluorometric assay to detect HIV-2 Protease activity.

  • Simple, rapid
  • Can detect HIV-2 protease activity in samples and of purified enzyme
  • Cleaves synthetic peptide substrates to release the free fluorophore

Human Immunodeficiency Virus (HIV) is the cause of the Acquired Immunodeficiency Syndrome (AIDS). Infection with HIV-2 virus causes immunosuppression, as well as AIDS, characterized by the same signs, symptoms and opportunistic infections as HIV-1. However, HIV-2-associated AIDS may often be associated with lower viral load levels than HIV-1. Aspartyl HIV-2 Protease (retropepsin) is an essential enzyme for the life-cycle of HIV virus. Expressed as a part of the Gag-Pol polyprotein complex in the infected cells, it is auto-catalytically released upon formation of an immature viral particle. The enzyme subsequently cleaves the other parts of viral polyproteins resulting in the maturation of the virus. In HIV-infected patients the enzyme is subjected to intensive mutagenesis and mutants resistant to applied medicines are produced as a result of the selection pressure. BioVision’s HIV-2 Protease Activity assay kit utilizes the ability of active HIV-2 Protease to cleave a synthetic peptide substrate to release the free fluorophore which can be easily quantified using a fluorometer or fluorescence microplate reader. This assay is simple, rapid and can detect HIV-2 protease activity in samples and of purified enzyme.




 

ORDER

Expand All / Collapse All
Description Size Storage Temperature Ex/Em Supplier No. VWR Catalog Number Unit Price Quantity
HIV-2 Protease Activity Assay Kit (Fluorometric) 100 assays -80°C 330 to 450 nm K845-100 75832-898 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Sample Type Proteins/lysates

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty