Triclosan ≥97% (by HPLC), Calbiochem®
|
Warning
|
Synonyms:
5-Chloro-2-(2, 4-dichlorophenoxy) phenol
, 5-Chloro-2-(2,4-dichlorophenoxy)phenol
Packaged under inert gas.
647950-1GM
CA80511-110
Triclosan ≥97% (by HPLC), Calbiochem®
Triclosan
A potent antimicrobial agent that inhibits bacterial fatty acid synthesis by selectively targeting Fabl encoded enoyl-acyl carrier protein (ACP) reductase. A slow-binding inhibitor that forms a stable ternary complex (bsFabl.NAD+.triclosan) in
Special Instructions: Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Formula:
C₁₂H₇Cl₃O₂ MW: 289.54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00800992 CAS Number: 3380-34-5 EINECS: 222-182-2 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility | DMSO (50 mg/mL), Ethanol (50 mg/mL) |
Purity by HPLC | 99.7% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...