Glycine Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB211100-100TEST
75832-870EA 1039.82 USD
75832-870
Glycine Assay Kit (Fluorometric), BioVision Inc.
Assays Protein Assays
Market's first and only kit for enzymatic detection of glycine in biological samples.
  • Measurement of glycine in biological samples such as serum, plasma, saliva, urine, etc.
  • Standard and non-standard amino acids do not interfere with the assay
  • Can detect as little as 1 µM of Glycine
Glycine (GLY) is one of the 20 standard amino acids commonly found in proteins. GLY side chain is a hydrogen substituent, thus it is the only amino acid that is not chiral and the smallest among the proteogenic amino acids. Basic functions of Glycine include the participation in the synthesis of creatine, glutathione, heme groups, and conjugated bile acids (bile salts). It is also present as one of the most abundant residues in the triple-helical structure of collagen. It acts as a glucogenic amino acid regulating sugar levels in blood. Therefore, glycine supplementation has been used in patients suffering anemia, hypoglycemia and chronic fatigue. In cancer cells, GLY consumption is highly correlated to cancer cell proliferation, via purine synthesis. Glycine uptake in cancer cell studies supports the role of this amino acid in tumorigenesis and malignancy. GLY possesses both inhibitory and excitatory neurotransmitter functions in the brain stem and spinal cord. BioVision’s Glycine Assay Kit provides a simple, sensitive, and high-throughput adaptable assay that detects physiological concentration of glycine in a variety of biological fluids. The principle of the assay is based on the oxidation of glycine producing a fluorophore (Ex/Em = 535/587 nm) with a stable signal, which is directly proportional to the amount of GLY in the sample. The assay is specific and other standard and non-standard amino acids do not interfere with the assay. The assay can detect as little as 1 µM of Glycine in a variety of samples.