PicoProbe™ ADP Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB211087-100TEST
75832-956EA 873.95 USD
75832-956
PicoProbe™ ADP Assay Kit (Fluorometric), BioVision Inc.
Assays Cellular Assays
Fast, sensitive, HTP compatible, fluorometric ADP assay with detection limit as low as 10 pmol or 0.05 µM of ADP.
  • Ultrasensitive, Simple, HTP adaptable fluoromotericassay kit for ADP measurement.
  • Can detect as low as 10 pmol or 0.05 µM of ADP Detection of NOS activity
Adenosine diphosphate (ADP) is an organic molecule that plays a critical role in cellular metabolism and energy transfer reactions. ADP is a product of ATP dephosphorylation and it can be rephosphorylated to ATP. These dephosphorylations and rephosphorylations occur via various phosphorylases and kinases. ADP is stored in platelets and can be released to interact with variety of purinergic receptors. ADP levels regulate several enzymes involved in intermediary metabolism. ADP conversion to ATP primarily occurs within the mitochondrion and chloroplast although several such processes also occur in the cytoplasm. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, the luciferase system is unstable and luminescence equipment is generally not available in most laboratories. BioVision’s PicoProbe™ ADP Assay Kit provides a sensitive fluorescence method to detect ADP. The kit is suitable for measuring ADP levels in various samples, including the ones that contain reducing substances, which may interfere with oxidase-based assays. In this assay, ADP, in the presence of ADP Enzyme Mix is converted to an intermediate, which reacts with the fluorescent probe to generate a strong stable signal at Ex/Em = 535/587 nm. BioVision’s PicoProbe™ ADP Assay kit provides an ultrasensitive, fast and high-throughput ready assay for detection of as low as 10 pmol or 0.05 µM of ADP in samples.