Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EZCell™ Intracellular Nitric Oxide Synthase (NOS) Detection Kit, BioVision Inc.

Supplier: BioVision
K207-100
Cell-based assay with a simple, one step, non-radiometric protocol for detection of intracellular NOS.

  • Detection of NOS activity in adherent cells
  • Screening, studying, and characterizing stimulators/inhibitors that affect intracellular levels of NOS
  • Simple, non-radiometric method

Nitric oxide synthases (EC 1.14.13.39) (NOSs) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. There are three isoforms of NOS: endothelial (eNOS), neuronal (nNOS), and inducible (iNOS). nNOS accounts for the production of NO in central nervous system, where NO participates in cell communication and information storage. eNOS produces NO in blood vessels and is involved with the regulation of vascular function. In contrast to other isoforms, iNOS is expressed de novo under oxidative stress conditions and produces large amounts of NO as a part of body’s defense mechanism. BioVision’s Intracellular Nitric Oxide Synthase Detection kit uses a dye that reacts with intracellular NO produced by NOS to produce fluorescence (Ex/Em = 485/530 nm), which is proportional to the concentration of intracellular NOS and can be detected using a microplate reader or a fluorescence microscope. This kit provides a simple, non-radiometric method for detection of intracellular NOS in the cells.




 

ORDER

Expand All / Collapse All
Description Size Storage Temperature Ex/Em Supplier No. VWR Catalog Number Unit Price Quantity
EZCell™ Intracellular Nitric Oxide Synthase (NOS) Detection Kit 100 assays -20°C 485 to 530 nm K207-100 75832-950 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Shelf Life 12 months
Species Reactivity Mammalian cells
Sample Type NOS producing cells or cell lines
Detection Method Fluorescence microscope

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty