BioWhittaker® BME Cell Culture Media, Lonza
Supplier: Lonza
12-132A 12-105F 12-105F 12-132A
12001-532EA 70.16 USD
CA12001-550 CA12001-532 12001-532 12001-550
BioWhittaker® BME Cell Culture Media, Lonza
Cell Culture Media
Basal medium eagle was formulated in 1955 for nutritional requirements of HeLa and mouse L cells
  • Culture primary or diploid mammalian cells
  • Suitable for a variety of cell types
  • Historical precursor to MEM
Basal medium eagle was formulated in 1955 for nutritional requirements of HeLa and mouse L cells. BME can be used for culturing primary or diploid mammalian cells. A minimal medium suitable for a variety of cell types. It is a historical precursor to Minimum Essential Medium (MEM).
All BioWhittaker™ Cell Culture Media products are manufactured in accordance with cGMP regulations. The formulations used for standard liquid media products are those recommended by the Tissue Culture Association.