Anti-AKT1 Rabbit Monoclonal Antibody [clone: RM252]
Supplier: Revmab Biosciences
31-1132-00
103255-336EA 382.31 USD
103255-336
Anti-AKT1 Rabbit Monoclonal Antibody [clone: RM252]
Antibodies
This antibody reacts to human Akt1. This antibody may also react to bovine, mouse or rat Akt1, as predicted by immunogen homology
Immunohistochemistry (IHC): 1:500-1:1000 dilution;
Western Blot (WB): 1:1000-1:2000 dilution.