Anti-ACTA2 Rabbit Monoclonal Antibody [clone: RM253]
Supplier: Revmab Biosciences
31-1133-00
103255-334EA 382.31 USD
103255-334
Anti-ACTA2 Rabbit Monoclonal Antibody [clone: RM253]
Antibodies
This antibody reacts to human and mouse alpha smooth muscle Actin (alpha-Actin-2). This antibody may also react to bovine or rat alpha smooth muscle Actin, as predicted by immunogen homology
Immunohistochemistry (IHC): 1:100-1:2500 dilution;
Western Blot (WB): 1:1000-1:2000 dilution.