Anti-GFAP Rabbit Monoclonal Antibody [clone: RM246]
Supplier: Revmab Biosciences
31-1127-00
103255-324EA 382.31 USD
103255-324
Anti-GFAP Rabbit Monoclonal Antibody [clone: RM246]
Antibodies
This antibody reacts to human and mouse GFAP (Glial fibrillary acidic protein).
Immunohistochemistry (IHC): 1:200-1:500 dilution;
Western Blot (WB): 1:1000-1:3000 dilution.