Anti-p38 MAPK Rabbit Monoclonal Antibody [clone: RM245]
Supplier: Revmab Biosciences
31-1126-00
103255-322EA 382.31 USD
103255-322
Anti-p38 MAPK Rabbit Monoclonal Antibody [clone: RM245]
Antibodies
This antibody reacts to human p38 MAPK. This antibody may also react to rat and mouse p38 MAPK, as predicted by immunogen homology.
Immunohistochemistry (IHC): 1:1000-1:5000 dilution;
Western Blot (WB): 1:1000-1:2000 dilution.