Anti-ACC1 Rabbit Monoclonal Antibody [clone: RM232]
Supplier: Revmab Biosciences
31-1116-00
103255-604EA 382.31 USD
103255-604
Anti-ACC1 Rabbit Monoclonal Antibody [clone: RM232]
Antibodies
This antibody reacts to human Acetyl CoA Carboxylase 1. This antibody may also react to mouse or rat Acetyl CoA Carboxylase 1, as predicted by immunogen homology.
Immunohistochemistry (IHC): 1:300-1:500 dilution;
Western Blot (WB): 1:1000-1:2000 dilution.