Anti-BRAF V600E Rabbit Monoclonal Antibody [clone: RM8]
Supplier: Revmab Biosciences
31-1042-00-S
103255-600EA 299 USD
103255-600
Anti-BRAF V600E Rabbit Monoclonal Antibody [clone: RM8]
Antibodies
This antibody reacts to the BRAF V600E mutant. No cross reactivity with wild type BRAF.
Immunohistochemistry (IHC): 2 – 10 ug/mL;
Immunocytochemistry (ICC): 2 – 10 ug/mL;
Western Blot (WB): 0.5 – 2 ug/mL