Anti-CK5 Rabbit Monoclonal Antibody [clone: RM226]
Supplier: Revmab Biosciences
31-1105-00-R
103255-582EA 686.17 USD
103255-582
Anti-CK5 Rabbit Monoclonal Antibody [clone: RM226]
Antibodies
This antibody reacts to Human Cytokeratin 5. This antibody may also react to bovine, mouse, or rat Cytokeratin, as predicted by immunogen homology.
Immunohistochemistry (IHC): 1:100 dilution;
Western Blot (WB): 1:1000 dilution.