Anti-His Tag Rabbit Monoclonal Antibody [clone: RM146]
Supplier: Revmab Biosciences
31-1048-00
103255-578EA 407.08 USD
103255-578
Anti-His Tag Rabbit Monoclonal Antibody [clone: RM146]
Antibodies
This antibody reacts to recombinant proteins containing the 6xHis-Tag or 10xHis-Tag fused to either the amino or carboxy terminus. No cross reactivity with other endogenous protein in mammalian or bacteria cells.
Western Blot: 0.1 ug/mL – 1 ug/mL;
Immunoprecipitation: 0.5 ug/mL – 2 ug/mL;
Immunocytochemistry 0.5 ug/mL – 2 ug/mL;
Flow Cytometry: 0.5 ug/mL – 2 ug/mL;
Immunohistochemistry; 0.1 ug/mL-1 ug/mL.