Anti-H2AX Rabbit Monoclonal Antibody [clone: RM214]
31-1090-00-S
103255-568EA 182 USD
103255-568
Anti-H2AX Rabbit Monoclonal Antibody [clone: RM214]
This antibody reacts to Histone H2AX protein, independent of post-translational modifications. No cross reactivity with other histone proteins. .
Western Blot: 0.5 ug/mL – 2 ug/mL;
ICC: 1 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.2 ug/mL – 1 ug/mL.