Anti-H4 Rabbit Monoclonal Antibody [clone: RM212]
31-1089-00
103255-560EA 429.69 USD
103255-560
Anti-H4 Rabbit Monoclonal Antibody [clone: RM212]
This antibody reacts to the Histone H4 protein, independent of post-translational modifications. No cross reactivity with other histone proteins.
Western Blot: 0.1 ug/mL – 0.5 ug/mL;
ICC: 1 ?g/mL - 2 ?g/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.