Anti-H2AZ Rabbit Monoclonal Antibody [clone: RM215]
Supplier: Revmab Biosciences
31-1091-00
103255-554EA 465.23 USD
103255-554
Anti-H2AZ Rabbit Monoclonal Antibody [clone: RM215]
Antibodies
This antibody reacts to Histone H2AZ protein, independent of post-translational modifications. No cross reactivity with other histone proteins.
Western Blot: 0.1 ug/mL – 0.5 ug/mL;
ICC: 1 ?g/mL - 2 ?g/mL;
ELISA: 0.1 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.