Anti-H2AX Rabbit Monoclonal Antibody [clone: RM214]
Supplier: Revmab Biosciences
31-1090-00
103255-552EA 465.23 USD
103255-552
Anti-H2AX Rabbit Monoclonal Antibody [clone: RM214]
Antibodies
This antibody reacts to Histone H2AX protein, independent of post-translational modifications. No cross reactivity with other histone proteins. .
Western Blot: 0.5 ug/mL – 2 ug/mL;
ICC: 1 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.2 ug/mL – 1 ug/mL.