Anti-H2B Rabbit Monoclonal Antibody [clone: RM230]
Supplier: Revmab Biosciences
31-1112-00
103255-550EA 474.92 USD
103255-550
Anti-H2B Rabbit Monoclonal Antibody [clone: RM230]
Antibodies
This antibody reacts to the Histone H2B protein, independent of post-translational modifications. No cross reactivity with other histone proteins.
Western Blot: 0.1 ug/mL – 1 ug/mL;
ICC: 0.5 ?g/mL - 1 ?g/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.2 ug/mL – 1 ug/mL.