Anti-H2A Rabbit Monoclonal Antibody [clone: RM225]
Supplier: Revmab Biosciences
31-1108-00
103255-548EA 465.23 USD
103255-548
Anti-H2A Rabbit Monoclonal Antibody [clone: RM225]
Antibodies
This antibody reacts to the Histone H2A protein, independent of post-translational modifications. No cross reactivity with other histone proteins.
Western Blot: 0.5 ug/mL – 2 ug/mL;
ICC: 1 ?g/mL - 2 ?g/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.2 ug/mL – 1 ug/mL.