Anti-H2AZK7ac Rabbit Monoclonal Antibody [clone: RM222]
31-1102-00-S
103255-298EA 182 USD
103255-298
Anti-H2AZK7ac Rabbit Monoclonal Antibody [clone: RM222]
This antibody reacts to Histone H2A.Z acetylated at Lysine 7 (K7ac). No cross reactivity with non-modified Lysine 7 or other acetylated Lysines in histone H2A.
Western Blot: 0.5 ug/mL – 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.05 ug/mL – 0.5 ug/mL.