Anti-H4K8ac Rabbit Monoclonal Antibody [clone: RM201]
31-1082-00-S
103255-268EA 182 USD
103255-268
Anti-H4K8ac Rabbit Monoclonal Antibody [clone: RM201]
This antibody reacts to Histone H4 acetylated at Lysine 8 (K8ac). No cross reactivity with other acetylated Lysines in Histone H4.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.05 ug/mL – 1 ug/mL.