Anti-H4S1p Rabbit Monoclonal Antibody [clone: RM194]
31-1079-00-S
103255-262EA 182 USD
103255-262
Anti-H4S1p Rabbit Monoclonal Antibody [clone: RM194]
This antibody reacts to Histone H4 phosphorylated at Serine 1. The reactivity is not affected by neighbor Arginine 3 modification. No cross reactivity with Histone H2A phosphorylated at Serine 1.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ICC: 1 ?g/mL - 2 ?g/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.