Anti-H3K23me2 Rabbit Monoclonal Antibody [clone: RM171]
31-1077-00-S
103255-256EA 182 USD
103255-256
Anti-H3K23me2 Rabbit Monoclonal Antibody [clone: RM171]
This antibody reacts to Histone H3 dimethylated at Lysine 23 (K23me2). No cross reactivity with monomethylated Lysine 23 (K23me1) or trimethylated Lysine 23 (K23me3), or other methylation in histone H3.
Western Blot: 0.5 ug/mL – 2 ug/mL;
ELISA: 0.1 ug/mL - 0.5 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.