Anti-H3K18me2 Rabbit Monoclonal Antibody [clone: RM168]
31-1075-00-S
103255-252EA 182 USD
103255-252
Anti-H3K18me2 Rabbit Monoclonal Antibody [clone: RM168]
This antibody reacts to Histone H3 dimethylated at Lysine 18 (K18me2). No cross reactivity with monomethylated Lysine 18 (K18me1) or trimethylated Lysine 18 (K18me3), or other methylation in histone H3.
Western Blot: 0.2 ug/mL – 1 ug/mL;
ELISA: 0.1 ug/mL - 0.5 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.