Anti-H3K27ac Rabbit Monoclonal Antibody [clone: RM172]
31-1056-00-S
103255-238EA 182 USD
103255-238
Anti-H3K27ac Rabbit Monoclonal Antibody [clone: RM172]
This antibody reacts to Histone H3 acetylated at Lysine 27 (K27ac). No cross reactivity with other acetylated Lysines in histone H3.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.