Anti-H2AZK4ac Rabbit Monoclonal Antibody [clone: RM221]
Supplier: Revmab Biosciences
31-1101-00
103255-522EA 465.23 USD
103255-522
Anti-H2AZK4ac Rabbit Monoclonal Antibody [clone: RM221]
Antibodies
This antibody reacts to Histone H2A.Z acetylated at Lysine 4 (K4ac). No cross reactivity with non-modified Lysine 4 or other acetylated Lysines in histone H2A
Western Blot: 0.5 ug/mL – 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.05 ug/mL – 0.5 ug/mL.