Anti-H4K20me3 Rabbit Monoclonal Antibody [clone: RM208]
Supplier: Revmab Biosciences
31-1096-00
103255-520EA 465.23 USD
103255-520
Anti-H4K20me3 Rabbit Monoclonal Antibody [clone: RM208]
Antibodies
This antibodyreacts to Histone H4 trimethylated at Lysine 20 (K20me3). It may slightly cross react to dimethylated Lysine 20 (K20me2) at a higher concentration. No cross reactivity with non-modified Lysine 20, monomethylated Lysine 20 (K20me1), or other methylations in histone H4.
Western Blot: 0.01 ug/mL - 1 ug/mL;
ELISA: 0.01 ug/mL - 0.5 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.