Anti-H3T6p Rabbit Monoclonal Antibody [clone: RM160]
Supplier: Revmab Biosciences
31-1093-00
103255-512EA 465.23 USD
103255-512
Anti-H3T6p Rabbit Monoclonal Antibody [clone: RM160]
Antibodies
This antibody reacts to Histone H3 phosphorylated at Threonine 6. No cross reactivity with other phosphorylated histones
Western Blot: 0.1 ug/mL - 1 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.