Anti-H3pSer10 Rabbit Monoclonal Antibody [clone: RM163]
Supplier: Revmab Biosciences
31-1070-00
103255-508EA 465.23 USD
103255-508
Anti-H3pSer10 Rabbit Monoclonal Antibody [clone: RM163]
Antibodies
This antibody reacts to Histone H3 phosphorylated at Serine 10. No cross reactivity with other phosphorylated histones.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ICC: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.