Anti-H2A/H4S1p Rabbit Monoclonal Antibody [clone: RM216]
Supplier: Revmab Biosciences
31-1092-00
103255-506EA 465.23 USD
103255-506
Anti-H2A/H4S1p Rabbit Monoclonal Antibody [clone: RM216]
Antibodies
This antibody reacts to Histone H4 or Histone H2A phosphorylated at Serine 1. No cross reactivity with other phosphorylated Histones.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ICC: 1 ?g/mL - 2 ?g/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.