Anti-H4K12ac Rabbit Monoclonal Antibody [clone: RM202]
Supplier: Revmab Biosciences
31-1088-00
103255-504EA 465.23 USD
103255-504
Anti-H4K12ac Rabbit Monoclonal Antibody [clone: RM202]
Antibodies
This antibody reacts to Histone H4 acetylated at Lysine 12 (K12ac). No cross reactivity with other acetylated Lysines in Histone H4.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.5 ug/mL - 1 ug/mL;
Multiplex: 0.5 ug/mL – 2 ug/mL.