Anti-H3K23ac Rabbit Monoclonal Antibody [clone: RM169]
Supplier: Revmab Biosciences
31-1087-00
103255-502EA 465.23 USD
103255-502
Anti-H3K23ac Rabbit Monoclonal Antibody [clone: RM169]
Antibodies
This antibody reacts to Histone H3 acetylated at Lysine 23 (K23ac). No cross reactivity with other acetylated Lysines in histone H3.
Western Blot: 0.5 ug/mL - 2 ug/mL;
ELISA: 0.5 ug/mL - 1 ug/mL;
Multiplex: 0.2 ug/mL – 1 ug/mL.