Anti-H3 Rabbit Monoclonal Antibody [clone: RM186]
Supplier: Revmab Biosciences
31-1086-00
103255-500EA 465.23 USD
103255-500
Anti-H3 Rabbit Monoclonal Antibody [clone: RM186]
Antibodies
This antibody recognizes Histone H3 that is unmodified at Lys4 but does not recognize acetylated or methylated Lys4. The antibody binding specificity allows for modifications of Arg2, Thr3, and/or other modifications in Histone H3.
Western Blot: 1 ug/mL – 2 ug/mL;
ELISA: 0.5 ug/mL - 1 ug/mL;
Multiplex: 0.5 ug/mL – 2 ug/mL.