Anti-H4K5ac Rabbit Monoclonal Antibody [clone: RM199]
Supplier: Revmab Biosciences
31-1081-00
103255-492EA 465.23 USD
103255-492
Anti-H4K5ac Rabbit Monoclonal Antibody [clone: RM199]
Antibodies
This antibody reacts to Histone H4 acetylated at Lysine 5 (K5ac). No cross reactivity with other acetylated Lysines in Histone H4.
Western Blot: 0.2 ug/mL - 1 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.05 ug/mL – 0.2 ug/mL.