Anti-H3K79me2 Rabbit Monoclonal Antibody [clone: RM181]
Supplier: Revmab Biosciences
31-1065-00
103255-484EA 465.23 USD
103255-484
Anti-H3K79me2 Rabbit Monoclonal Antibody [clone: RM181]
Antibodies
This antibody reacts to Histone H3 dimethylated at Lysine 79 (K79me2). No cross reactivity with monomethylated Lysine 79(K79me1) or trimethylated Lysine 79 (K79me3), or other methylation in histone H3.
Western Blot: 0.25 ug/mL – 1 ug/mL;
IHC: 0.1 ug/mL – 1 ug/mL;
ChIP: 2 ug/mL- 10 ug.mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.