Anti-H3K23me2 Rabbit Monoclonal Antibody [clone: RM171]
Supplier: Revmab Biosciences
31-1077-00
103255-482EA 465.23 USD
103255-482
Anti-H3K23me2 Rabbit Monoclonal Antibody [clone: RM171]
Antibodies
This antibody reacts to Histone H3 dimethylated at Lysine 23 (K23me2). No cross reactivity with monomethylated Lysine 23 (K23me1) or trimethylated Lysine 23 (K23me3), or other methylation in histone H3.
Western Blot: 0.5 ug/mL – 2 ug/mL;
ELISA: 0.1 ug/mL - 0.5 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.